Gene Transcripts
- Gene Symbol
- HIVEP1
- Entrez Gene ID
- 3096
- Gene Name
- human immunodeficiency virus type I enhancer binding protein 1
- Gene Aliases
- CIRIP, CRYBP1, GAAP, MBP-1, PRDII-BF1, Schnurri-1, ZAS1, ZNF40, ZNF40A
- Chromosome Location
- Chr. 6: 12002096 - 12212034 on Build GRCh38
- UniGene ID
- Hs.567284
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000607445.1 |
- |
2 |
252 |
|
Chr. 6 - 12007870 - 12008643 [+] |
View Full Product Details