Gene Transcripts

Gene Symbol
MAPKAPK5-AS1
Entrez Gene ID
51275
Gene Name
MAPKAPK5 antisense RNA 1
Gene Aliases
C12orf47
Chromosome Location
Chr. 12: 111839769 - 111842902 on Build GRCh38
UniGene ID
Hs.333120
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_015404.1 1 1102
GenBank mRNA AK096935.1 - 1 1102
AK124158.1 - 1 123
BC007973.2 - 1 106
BC015633.2 - 1 118
BC023547.2 - 1 753
BC107695.1 - 1 147
BC136540.1 - 1 1021
JQ794500.1 - 1 1205


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GENCODE ENST00000442119.5 - 1 106 Chr. 12 - 111839770 - 111841906 [-]
GENCODE ENST00000428207.4 - 1 1102 Chr. 12 - 111839769 - 111842902 [-]
GENCODE ENST00000443596.1 - 1 102 Chr. 12 - 111839767 - 111841902 [-]
GENCODE ENST00000456429.5 - 1 1102 Chr. 12 - 111839768 - 111842902 [-]

View Full Product Details