Gene Transcripts
- Gene Symbol
- LOC101928567
- Entrez Gene ID
- 101928567
- Gene Name
- uncharacterized LOC101928567
- Gene Aliases
- Chromosome Location
- Chr. 17: 15806242 - 15817742 on Build GRCh38
- UniGene ID
- Hs.569757
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_110839.1 |
|
3 |
673 |
|
- Gene Symbol
- UBE2R2
- Entrez Gene ID
- 54926
- Gene Name
- ubiquitin conjugating enzyme E2 R2
- Gene Aliases
- CDC34B, E2-CDC34B, UBC3B
- Chromosome Location
- Chr. 9: 33817164 - 33920403 on Build GRCh38
- UniGene ID
- Hs.643648
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC038356.1 |
- |
|
591 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000418821.2 |
- |
3 |
673 |
|
Chr. 17 - 15806241 - 15817742 [-] |
View Full Product Details