Gene Transcripts
- Gene Symbol
- STX6
- Entrez Gene ID
- 10228
- Gene Name
- syntaxin 6
- Gene Aliases
- Chromosome Location
- Chr. 1: 180949690 - 181023121 on Build GRCh38
- UniGene ID
- Hs.518417
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK056657.1 |
- |
2 |
2176 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000358073.2 |
- |
2 |
2176 |
|
Chr. 1 - 180944039 - 180976482 [-] |
View Full Product Details