Gene Transcripts

Gene Symbol
GNPNAT1
Entrez Gene ID
64841
Gene Name
glucosamine-phosphate N-acetyltransferase 1
Gene Aliases
GNA1, GNPNAT, Gpnat1
Chromosome Location
Chr. 14: 52775193 - 52791668 on Build GRCh38
UniGene ID
Hs.478025
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA CA435370.1 - 1 453


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GENCODE ENST00000555969.1 - 1 392 Chr. 14 - 52775237 - 52777740 [+]

View Full Product Details