Gene Transcripts

Gene Symbol
NR4A1
Entrez Gene ID
3164
Gene Name
nuclear receptor subfamily 4 group A member 1
Gene Aliases
GFRP1, HMR, N10, NAK-1, NGFIB, NP10, NUR77, TR3
Chromosome Location
Chr. 12: 52022832 - 52059507 on Build GRCh38
UniGene ID
Hs.524430
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BG057763.1 - 1 329


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA HY261392.1 - 1 341 Chr. 12 - 52059046 - 52059503 [-]
GenBankmRNA HY273074.1 - 1 340 Chr. 12 - 52059114 - 52059503 [-]
GENCODE ENST00000564363.1 - 1 312 Chr. 12 - 52058750 - 52059503 [-]
GenBankmRNA HY264705.1 - 1 345 Chr. 12 - 52059138 - 52059509 [-]
GenBankmRNA HY284348.1 - 1 361 Chr. 12 - 52059073 - 52059503 [-]
GenBankmRNA HY294431.1 - 1 339 Chr. 12 - 52059100 - 52059503 [-]
GenBankmRNA HY248926.1 - 1 338 Chr. 12 - 52059070 - 52059503 [-]

View Full Product Details