Gene Transcripts
- Gene Symbol
- DTX2P1-UPK3BP1-PMS2P11
- Entrez Gene ID
- 441263
- Gene Name
- DTX2P1-UPK3BP1-PMS2P11 readthrough, transcribed pseudogene
- Gene Aliases
- PMS2L11
- Chromosome Location
- Chr. 7: 76980822 - 77023761 on Build GRCh38
- UniGene ID
- Hs.675888
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_023383.1 |
|
11 |
1750 |
|
- Gene Symbol
- PMS2P2
- Entrez Gene ID
- 5380
- Gene Name
- PMS1 homolog 2, mismatch repair system component pseudogene 2
- Gene Aliases
- PMS2L2, PMS4
- Chromosome Location
- Chr. 7: 75343937 - 75358997 on Build GRCh38
- UniGene ID
- Hs.292996
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003614.3 |
|
5 |
622 |
|
- Gene Symbol
- PMS2P4
- Entrez Gene ID
- 5382
- Gene Name
- PMS1 homolog 2, mismatch repair system component pseudogene 4
- Gene Aliases
- PMS2L4, PMS6
- Chromosome Location
- Chr. 7: 67276131 - 67302442 on Build GRCh38
- UniGene ID
- Hs.712714
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_046297.1 |
|
6 |
728 |
|
- Gene Symbol
- PMS2P5
- Entrez Gene ID
- 5383
- Gene Name
- PMS1 homolog 2, mismatch repair system component pseudogene 5
- Gene Aliases
- PMS2L5, PMS7
- Chromosome Location
- Chr. 7: 74890768 - 74921138 on Build GRCh38
- UniGene ID
- Hs.292996
- Species
- Homo sapiens
- Gene Symbol
- PMS2P9
- Entrez Gene ID
- 100132832
- Gene Name
- PMS1 homolog 2, mismatch repair system component pseudogene 9
- Gene Aliases
- PMS2L17, PMS2LP1, PMSR5
- Chromosome Location
- Chr. 7: 77039480 - 77053038 on Build GRCh38
- UniGene ID
- Hs.292996
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_028058.1 |
|
5 |
1134 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000584900.5 |
- |
11 |
1623 |
|
Chr. 7 - 76980949 - 77023761 [+] |
View Full Product Details