Gene Transcripts
- Gene Symbol
- FLT3LG
- Entrez Gene ID
- 2323
- Gene Name
- fms related tyrosine kinase 3 ligand
- Gene Aliases
- FL, FLT3L
- Chromosome Location
- Chr. 19: 49474172 - 49487038 on Build GRCh38
- UniGene ID
- Hs.428
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK301136.1 |
- |
8 |
1096 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000595815.1 |
- |
8 |
1096 |
|
Chr. 19 - 49474586 - 49487637 [+] |
View Full Product Details