Gene Transcripts

Gene Symbol
Gas5
Entrez Gene ID
14455
Gene Name
growth arrest specific 5
Gene Aliases
Gas-5, Mir5117
Chromosome Location
Chr. 1: 161035166 - 161038537 on Build GRCm38
UniGene ID
Mm.270065
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_002840.2 1, 2 2390
GenBank mRNA AK083453.1 - 1, 2 2391
AK206779.1 - 9, 10 321
AK219294.1 - 9, 10 321
BC055883.1 - 5, 6 901
BC080708.1 - 3, 4 1447


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DV061968.1 - 7, 8 642 Chr. 1 - 161035164 - 161038534 [+]
GenBankmRNA DV063492.1 - 339 Chr. - - [-]
GenBankmRNA BF661484.1 - 1, 2 241 Chr. 1 - 161037315 - 161038126 [+]

View Full Product Details