Gene Transcripts

Gene Symbol
Adh1
Entrez Gene ID
11522
Gene Name
alcohol dehydrogenase 1 (class I)
Gene Aliases
ADH-AA, AI194826, Adh-1, Adh-1-t, Adh-1e, Adh-1t, Adh-3e, Adh1-e, Adh1-t, Adh1tl, Adh3-e
Chromosome Location
Chr. 3: 138277645 - 138290691 on Build GRCm38
UniGene ID
Mm.2409
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK082149.1 - 947

View Full Product Details