Gene Transcripts

Gene Symbol
Rmst
Entrez Gene ID
110333
Gene Name
rhabdomyosarcoma 2 associated transcript (non-coding RNA)
Gene Aliases
AI853140, C230053E11Rik, D930049J19Rik, Dmt2, M2, Ncrms
Chromosome Location
Chr. 10: 92081746 - 92165178 on Build GRCm38
UniGene ID
Mm.429764
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_028262.1 2 129
GenBank mRNA AK049726.1 - 2 94
AK082467.1 - 2 122
AK086758.1 - 2 122
AK136927.1 - 1 25
BB621570.1 - 2 130


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA CJ162880.1 - 2 122 Chr. 10 - 92135743 - 92165169 [-]
GenBankmRNA CJ182976.1 - 2 122 Chr. 10 - 92164420 - 92165169 [-]
GenBankmRNA AI255164.1 - 2 111 Chr. 10 - 92135751 - 92165160 [-]
GenBankmRNA BB652773.1 - 2 91 Chr. 10 - 92162090 - 92165140 [-]
GenBankmRNA AI255186.1 - 2 111 Chr. 10 - 92135751 - 92165160 [-]
GenBankmRNA BB663622.1 - 2 122 Chr. 10 - 92134963 - 92165170 [-]
GenBankmRNA CJ058684.1 - 1 28 Chr. 10 - 92135701 - 92164751 [-]
GenBankmRNA CJ062038.1 - 2 122 Chr. 10 - 92136560 - 92165170 [-]
GenBankmRNA CJ059102.1 - 2 122 Chr. 10 - 92135791 - 92165170 [-]
GenBankmRNA BX528278.1 - 2 112 Chr. 10 - 92135701 - 92165160 [-]
GenBankmRNA CJ159784.1 - 1 52 Chr. 10 - 92135702 - 92164776 [-]
GenBankmRNA CJ177102.1 - 1 52 Chr. 10 - 92135070 - 92164776 [-]
GenBankmRNA CJ057229.1 - 1 25 Chr. 10 - 92135037 - 92164748 [-]
GenBankmRNA CO425135.1 - 1 15 Chr. 10 - 92133279 - 92164739 [-]
GenBankmRNA CJ057228.1 - 1 25 Chr. 10 - 92135035 - 92164748 [-]
GenBankmRNA BB425109.2 - 2 104 Chr. 10 - 92162107 - 92165144 [-]
GenBankmRNA BY721509.1 - 1 22 Chr. 10 - 92134204 - 92164746 [-]
GenBankmRNA BB651040.1 - 2 122 Chr. 10 - 92164173 - 92165170 [-]
GenBankmRNA CJ173535.1 - 1 52 Chr. 10 - 92135701 - 92164776 [-]
GenBankmRNA CJ160605.1 - 2 122 Chr. 10 - 92135768 - 92165169 [-]
GenBankmRNA CJ069979.1 - 2 122 Chr. 10 - 92162107 - 92165170 [-]

View Full Product Details