Gene Transcripts
- Gene Symbol
- Mfn2
- Entrez Gene ID
- 170731
- Gene Name
- mitofusin 2
- Gene Aliases
- D630023P19Rik, Fzo
- Chromosome Location
- Chr. 4: 147873586 - 147904909 on Build GRCm38
- UniGene ID
- Mm.154312
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001285920.1 |
NP_001272849.1
|
12, 13 |
1645 |
|
NM_001285921.1 |
NP_001272850.1
|
12, 13 |
1628 |
|
NM_001285922.1 |
NP_001272851.1
|
11, 12 |
1559 |
|
NM_001285923.1 |
NP_001272852.1
|
11, 12 |
1559 |
|
NM_133201.3 |
NP_573464.2
|
11, 12 |
1562 |
|
GenBank mRNA |
AK042080.1 |
- |
11, 12 |
1343 |
|
AK049583.1 |
- |
11, 12 |
1352 |
|
AK052689.1 |
- |
11, 12 |
1343 |
|
AK122220.1 |
- |
10, 11 |
2537 |
|
AK146801.1 |
- |
12, 13 |
1405 |
|
AK147863.1 |
- |
11, 12 |
1358 |
|
AK169373.1 |
- |
11, 12 |
1347 |
|
AY123975.1 |
- |
11, 12 |
1509 |
|
BC046503.1 |
- |
11, 12 |
1340 |
4191179 |
View Full Product Details