Gene Transcripts

Gene Symbol
Klra17
Entrez Gene ID
170733
Gene Name
killer cell lectin-like receptor, subfamily A, member 17
Gene Aliases
Ly49q, Ly49q1
Chromosome Location
Chr. 6: 129831154 - 129876672 on Build GRCm38
UniGene ID
Mm.155514
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133203.5 NP_573466.3 4 428
GenBank mRNA AB033769.1 - 4 441
AB190787.1 - 3 287
AK080021.1 - 4 429
BC132236.1 - 4 389 40130513

View Full Product Details