Gene Transcripts
- Gene Symbol
- Vmn1r9
- Entrez Gene ID
- 171203
- Gene Name
- vomeronasal 1 receptor 9
- Gene Aliases
- V1rc30
- Chromosome Location
- Chr. 6: 57070895 - 57071945 on Build GRCm38
- UniGene ID
- Mm.377181
- Species
- Mus musculus
- Gene Symbol
- Vmn1r10
- Entrez Gene ID
- 113858
- Gene Name
- vomeronasal 1 receptor 10
- Gene Aliases
- V1rc1
- Chromosome Location
- Chr. 6: 57113425 - 57114360 on Build GRCm38
- UniGene ID
- Mm.436285
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_053231.2 |
NP_444461.2
|
1 |
604 |
|
View Full Product Details