Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_138665.2 | NP_619606.1 | 16 | 2699 | |
GenBank mRNA | AK087163.1 | - | 8 | 886 | |
AK140451.1 | - | 15 | 1976 | ||
AK159236.1 | - | 16 | 2663 | ||
AK159681.1 | - | 16 | 2663 | ||
AK159704.1 | - | 16 | 2663 | ||
AK163515.1 | - | 16 | 2228 | ||
AK163660.1 | - | 15 | 1975 | ||
BC003456.1 | - | 16 | 2208 | 2648344 |