Gene Transcripts

Gene Symbol
Fbxw10
Entrez Gene ID
213980
Gene Name
F-box and WD-40 domain protein 10
Gene Aliases
Fbw10, HREP, SM25H2, SM2SH2
Chromosome Location
Chr. 11: 62846990 - 62877464 on Build GRCm38
UniGene ID
Mm.482259
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033669.2 NP_001028841.2 8 1702
NM_001291441.1 NP_001278370.1 8 1702
GenBank mRNA AK132680.1 - 8 1703
BC141212.1 - 8 1693 9055824

View Full Product Details