Gene Transcripts

Gene Symbol
Pstk
Entrez Gene ID
214580
Gene Name
phosphoseryl-tRNA kinase
Gene Aliases
5430423O14Rik, 5730458D16Rik, Gm1099, R75284
Chromosome Location
Chr. 7: 131371113 - 131392768 on Build GRCm38
UniGene ID
Mm.272948
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039534.1 NP_001034623.1 2, 3 537
GenBank mRNA AK077585.1 - 2, 3 538
BC110381.1 - 2, 3 513 5149569
BC115503.2 - 2, 3 538

View Full Product Details