Gene Transcripts

Gene Symbol
Cdh12
Entrez Gene ID
215654
Gene Name
cadherin 12
Gene Aliases
Cdhb
Chromosome Location
Chr. 15: 20448879 - 21597005 on Build GRCm38
UniGene ID
Mm.300909
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008420.2 NP_001008420.1 11 1852
GenBank mRNA BC086655.1 - 11 1852

View Full Product Details