Gene Transcripts

Gene Symbol
Myl6b
Entrez Gene ID
216459
Gene Name
myosin, light polypeptide 6B
Gene Aliases
5730437E04Rik, BC037527, MLC1SA
Chromosome Location
Chr. 10: 128494157 - 128498685 on Build GRCm38
UniGene ID
Mm.291688
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172259.1 NP_758463.1 5 463
GenBank mRNA BC037527.1 - 5 463

View Full Product Details