Gene Transcripts

Gene Symbol
Fnip1
Entrez Gene ID
216742
Gene Name
folliculin interacting protein 1
Gene Aliases
A730024A03Rik, AI838773, AW557298
Chromosome Location
Chr. 11: 54438146 - 54518241 on Build GRCm38
UniGene ID
Mm.34087
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173753.4 NP_776114.2 17 3438
GenBank mRNA AK042779.1 - 4 903
AK047814.1 - 4 679
AK122574.1 - 17 3386
BC024776.1 - 2 285
BC079892.1 - 17 3428 30543296

View Full Product Details