Gene Transcripts

Gene Symbol
Stxbp6
Entrez Gene ID
217517
Gene Name
syntaxin binding protein 6 (amisyn)
Gene Aliases
BC024598, C85317
Chromosome Location
Chr. 12: 44852484 - 45074737 on Build GRCm38
UniGene ID
Mm.285400
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144552.3 NP_653135.2 2 455
GenBank mRNA AK158779.1 - 2 341
BC024598.1 - 2 337 4020199

View Full Product Details