Gene Transcripts

Gene Symbol
Wdhd1
Entrez Gene ID
218973
Gene Name
WD repeat and HMG-box DNA binding protein 1
Gene Aliases
A630092E18, AND-1, D630024B06Rik
Chromosome Location
Chr. 14: 47240944 - 47276857 on Build GRCm38
UniGene ID
Mm.265615
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172598.3 NP_766186.2 12 1494
GenBank mRNA AK036390.1 - 12 1564
AK052690.1 - 12 1581
BC063740.1 - 12 1427 6328626

View Full Product Details