Gene Transcripts

Gene Symbol
Phf11d
Entrez Gene ID
219132
Gene Name
PHD finger protein 11D
Gene Aliases
D14Ertd668e, Phf11, Phf11l
Chromosome Location
Chr. 14: 59347407 - 59370520 on Build GRCm38
UniGene ID
Mm.479285
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_199015.4 NP_950180.3 3 290
GenBank mRNA AK156583.1 - 3 271
BC030186.1 - 3 291 5067268
BC110663.1 - 3 247
BC145673.1 - 3 210 40130976
BG083212.2 - 3 290

View Full Product Details