Gene Transcripts

Gene Symbol
Rnd1
Entrez Gene ID
223881
Gene Name
Rho family GTPase 1
Gene Aliases
A830014L09Rik, Arhs
Chromosome Location
Chr. 15: 98669205 - 98677461 on Build GRCm38
UniGene ID
Mm.274010
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172612.3 NP_766200.1 3 339
GenBank mRNA AK039992.1 - 338
AK043639.1 - 3 340
AK161870.1 - 3 336
BC048531.1 - 3 336 6535763

View Full Product Details