Gene Transcripts
- Gene Symbol
- Cd226
- Entrez Gene ID
- 225825
- Gene Name
- CD226 antigen
- Gene Aliases
- BC051526, DNAM-1, DNAM1, Pta1, TLiSA1
- Chromosome Location
- Chr. 18: 89176954 - 89272232 on Build GRCm38
- UniGene ID
- Mm.139293
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001039149.1 |
NP_001034238.1
|
3, 4 |
628 |
|
NM_178687.2 |
NP_848802.2
|
4, 5 |
967 |
|
GenBank mRNA |
AB195429.1 |
- |
3, 4 |
740 |
|
AF416980.1 |
- |
4, 5 |
799 |
|
AF421199.1 |
- |
3, 4 |
460 |
|
AF421200.1 |
- |
2, 3 |
388 |
|
AK036455.1 |
- |
5, 6 |
1062 |
|
AK140349.1 |
- |
4, 5 |
855 |
|
BC132422.1 |
- |
4, 5 |
912 |
40130699 |
BC138769.1 |
- |
4, 5 |
912 |
8861791 |
BC145410.1 |
- |
3, 4 |
573 |
9053968 |
View Full Product Details