Gene Transcripts

Gene Symbol
Fras1
Entrez Gene ID
231470
Gene Name
Fraser syndrome 1 homolog (human)
Gene Aliases
E130113P14Rik, bl, mKIAA1500
Chromosome Location
Chr. 5: 96373825 - 96784732 on Build GRCm38
UniGene ID
Mm.291120
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175473.3 NP_780682.3 57, 58 9448
GenBank mRNA AJ489280.1 - 57, 58 9448
AK051850.1 - 3, 4 656
AK122518.1 - 10, 11 1745

View Full Product Details