Gene Transcripts

Gene Symbol
Rpap2
Entrez Gene ID
231571
Gene Name
RNA polymerase II associated protein 2
Gene Aliases
AW060207
Chromosome Location
Chr. 5: 107597373 - 107661853 on Build GRCm38
UniGene ID
Mm.482420
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163461.2 NP_001156933.1 8 1074
NM_001163462.2 NP_001156934.1 8 1131
NM_001289569.1 NP_001276498.1 8 1088
NM_001289570.1 NP_001276499.1 8 1074
NM_144911.3 NP_659160.2 8 1074
GenBank mRNA AK034418.1 - 8 1090
AK049889.1 - 8 1131
AK145830.1 - 8 1049
AK161901.1 - 8 1055
AK166574.1 - 8 997
BC021895.1 - 8 1035 3481916

View Full Product Details