Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001013441.2 | NP_001013459.1 | 3 | 472 | |
NM_183172.2 | NP_898995.1 | 3 | 472 | ||
GenBank mRNA | AK132369.1 | - | 3 | 376 | |
AK137531.1 | - | 2 | 270 | ||
AY940666.1 | - | 3 | 441 | ||
AY940667.1 | - | 3 | 429 | ||
BC051080.1 | - | 3 | 469 | 6490136 |