Gene Transcripts

Gene Symbol
Gpr75
Entrez Gene ID
237716
Gene Name
G protein-coupled receptor 75
Gene Aliases
Chromosome Location
Chr. 11: 30884600 - 30893729 on Build GRCm38
UniGene ID
Mm.338361
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175490.4 NP_780699.2 2 1120
GenBank mRNA AK044553.1 - 1 587
AK141614.1 - 2 1121
AY253852.1 - 1 827
BC125614.1 - 1 916 40130129
BC125616.1 - 1 916 40130131

View Full Product Details