Gene Transcripts

Gene Symbol
Sdk2
Entrez Gene ID
237979
Gene Name
sidekick homolog 2 (chicken)
Gene Aliases
4632412F08Rik, 5330435L01Rik, Sdk-2, mKIAA1514
Chromosome Location
Chr. 11: 113776374 - 114066181 on Build GRCm38
UniGene ID
Mm.130504
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172800.2 NP_766388.2 41 5699
GenBank mRNA AK129379.1 - 37 5080
AK161326.1 - 9 1065
AY351699.1 - 41 5699

View Full Product Details