Gene Transcripts

Gene Symbol
Cntnap3
Entrez Gene ID
238680
Gene Name
contactin associated protein-like 3
Gene Aliases
Chromosome Location
Chr. 13: 64736806 - 64904028 on Build GRCm38
UniGene ID
Mm.390590
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081129.1 NP_001074598.1 22 3622
GenBank mRNA AK141609.1 - 1245

View Full Product Details