Gene Transcripts

Gene Symbol
Plekhm3
Entrez Gene ID
241075
Gene Name
pleckstrin homology domain containing, family M, member 3
Gene Aliases
9430067K14Rik, A230102O09Rik, AI449570, DAPR, Plekhm1l
Chromosome Location
Chr. 1: 64789120 - 64956824 on Build GRCm38
UniGene ID
Mm.239534
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039493.1 NP_001034582.1 6 2268
GenBank mRNA AK034959.1 - 6 2190
AK172220.1 - 5 1361
BC063098.1 - 2268

View Full Product Details