Gene Transcripts

Gene Symbol
Fbxl5
Entrez Gene ID
242960
Gene Name
F-box and leucine-rich repeat protein 5
Gene Aliases
Fbl4, Fir4
Chromosome Location
Chr. 5: 43744618 - 43782149 on Build GRCm38
UniGene ID
Mm.25794
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001159963.1 NP_001153435.1 6 973
NM_178729.4 NP_848844.1 6 973
GenBank mRNA AK029957.1 - 6 921
AK041497.1 - 6 945
AK054009.1 - 5 3312
AK082931.1 - 6 943
AK085100.1 - 6 946
AK088067.1 - 6 940
AK150664.1 - 6 933
AK159744.1 - 6 962
AK171361.1 - 6 927
AK171783.1 - 6 833
AK177606.1 - 2 286
AK183837.1 - 2 286
BC023291.1 - 4 578
BC047214.1 - 886 6466511
BC113798.1 - 6 891 40062506

View Full Product Details