Gene Transcripts

Gene Symbol
Emilin2
Entrez Gene ID
246707
Gene Name
elastin microfibril interfacer 2
Gene Aliases
FOAP-10
Chromosome Location
Chr. 17: 71252172 - 71311639 on Build GRCm38
UniGene ID
Mm.23462
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145158.3 NP_660140.1 2 268
GenBank mRNA AF468645.1 - 2 219
AK154230.1 - 2 238
AK155037.1 - 2 246
AK155306.1 - 2 262
AK155366.1 - 2 249
AK155554.1 - 2 264
AK155797.1 - 2 265
AK155878.1 - 2 266
AK155879.1 - 2 262
AK155909.1 - 2 268
AK155913.1 - 2 238
AK155918.1 - 2 295
AK170081.1 - 2 265
AK170288.1 - 2 238
AK170391.1 - 2 238
AK170801.1 - 2 269
AK170822.1 - 2 290
AK170953.1 - 2 284
AK171015.1 - 2 290
AK171020.1 - 2 256
AK171303.1 - 2 240
AK171309.1 - 2 265
AK171579.1 - 2 238
AK171592.1 - 2 238
AK171616.1 - 2 238
BC053753.1 - 2 552 6510220
GQ872457.1 - 2 163
GQ872460.1 - 2 163

View Full Product Details