Gene Transcripts
- Gene Symbol
- MAP2K3
- Entrez Gene ID
- 5606
- Gene Name
- mitogen-activated protein kinase kinase 3
- Gene Aliases
- MAPKK3, MEK3, MKK3, PRKMK3, SAPKK-2, SAPKK2
- Chromosome Location
- Chr. 17: 21284656 - 21315240 on Build GRCh38
- UniGene ID
- Hs.514012
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001316332.1 |
NP_001303261.1
|
4, 5 |
559 |
|
NM_002756.4 |
NP_002747.2
|
3, 4 |
176 |
|
NM_145109.2 |
NP_659731.1
|
3, 4 |
434 |
|
XM_005256723.2 |
XP_005256780.1
|
4, 5 |
474 |
|
XM_011523958.2 |
XP_011522260.1
|
3, 4 |
233 |
|
XM_011523959.1 |
XP_011522261.1
|
5, 6 |
441 |
|
XM_017024857.1 |
XP_016880346.1
|
3, 4 |
303 |
|
XM_017024858.1 |
XP_016880347.1
|
4, 5 |
291 |
|
XM_017024859.1 |
XP_016880348.1
|
5, 6 |
631 |
|
GenBank mRNA |
AB451226.1 |
- |
3, 4 |
169 |
|
AB451349.1 |
- |
3, 4 |
169 |
|
AK093838.1 |
- |
4, 5 |
478 |
|
AK223490.1 |
- |
3, 4 |
309 |
|
AK315505.1 |
- |
3, 4 |
379 |
|
BC032478.1 |
- |
3, 4 |
369 |
5215093 |
BM836867.1 |
- |
3, 4 |
414 |
|
CR536514.1 |
- |
3, 4 |
169 |
|
CR541793.1 |
- |
3, 4 |
169 |
|
D87116.1 |
- |
3, 4 |
226 |
|
DA131175.1 |
- |
3, 4 |
434 |
|
DA987047.1 |
- |
3, 4 |
177 |
|
GQ225578.1 |
- |
3, 4 |
169 |
|
L36719.1 |
- |
5, 6 |
419 |
|
U66839.1 |
- |
3, 4 |
295 |
|
View Full Product Details