Gene Transcripts

Gene Symbol
Robo2
Entrez Gene ID
268902
Gene Name
roundabout homolog 2 (Drosophila)
Gene Aliases
2600013A04Rik, 9430089E08Rik, BB097918, D230004I22Rik, mKIAA1568
Chromosome Location
Chr. 16: 73892061 - 75447298 on Build GRCm38
UniGene ID
Mm.171736
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175549.4 NP_780758.3 2 317
GenBank mRNA AK031495.1 - 2 843
AK035110.1 - 2 1191
AK048081.1 - 2 1231
AK051227.1 - 2 1231
AK085461.1 - 2 942
AK164874.1 - 2 941
BC055333.1 - 2 625 6409283
DQ533876.1 - 2 317

View Full Product Details