Gene Transcripts

Gene Symbol
Lhfpl4
Entrez Gene ID
269788
Gene Name
lipoma HMGIC fusion partner-like protein 4
Gene Aliases
1190004M23Rik, AI604880, B230384L07, mKIAA4027
Chromosome Location
Chr. 6: 113168087 - 113195384 on Build GRCm38
UniGene ID
Mm.37943
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177763.3 NP_808431.2 3, 4 900
GenBank mRNA AK046437.1 - 3, 4 900
AK220449.1 - 3, 4 844
BC085130.1 - 3, 4 847 5704741

View Full Product Details