Gene Transcripts

Gene Symbol
Slc35e1
Entrez Gene ID
270066
Gene Name
solute carrier family 35, member E1
Gene Aliases
6030458H05, AA408278
Chromosome Location
Chr. 8: 72477995 - 72492614 on Build GRCm38
UniGene ID
Mm.291461
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177766.3 NP_808434.2 5, 6 1037
GenBank mRNA AK031600.1 - 5, 6 785
AK200930.1 - 2, 3 300
AK203664.1 - 1, 2 303
BC115553.1 - 5, 6 696 40101801
BC115554.1 - 5, 6 696 40101802

View Full Product Details