Gene Transcripts

Gene Symbol
Nlrx1
Entrez Gene ID
270151
Gene Name
NLR family member X1
Gene Aliases
BC034204, NOD9
Chromosome Location
Chr. 9: 44252713 - 44268599 on Build GRCm38
UniGene ID
Mm.3957
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163742.1 NP_001157214.1 3, 4 300
NM_001163743.1 NP_001157215.1 3, 4 401
NM_178420.3 NP_848507.2 3, 4 389
GenBank mRNA AK089260.1 - 3, 4 393
AK141296.1 - 3, 4 370
AK145780.1 - 3, 4 410
AK145988.1 - 3, 4 390
AK166689.1 - 3, 4 227
AK170088.1 - 3, 4 344
BC034204.1 - 2, 3 183
BC050054.1 - 3, 4 296 30012053
BY196718.1 - 3, 4 300

View Full Product Details