Gene Transcripts

Gene Symbol
Gimap9
Entrez Gene ID
317758
Gene Name
GTPase, IMAP family member 9
Gene Aliases
A630002K24, BB145400
Chromosome Location
Chr. 6: 48676121 - 48679116 on Build GRCm38
UniGene ID
Mm.116971
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_174960.2 NP_777620.1 2 872
GenBank mRNA BC038043.1 - 2 794
BC096680.1 - 2 856 1345477

View Full Product Details