Gene Transcripts

Gene Symbol
Usp37
Entrez Gene ID
319651
Gene Name
ubiquitin specific peptidase 37
Gene Aliases
4932415L06Rik, C330008N13Rik
Chromosome Location
Chr. 1: 74435510 - 74544286 on Build GRCm38
UniGene ID
Mm.471601
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_176972.4 NP_795946.2 17, 18 2214
GenBank mRNA AK030013.1 - 17, 18 2215
BC114208.1 - 15, 16 4329
BC139091.1 - 17, 18 2156 8862113

View Full Product Details