Gene Transcripts

Gene Symbol
Eml5
Entrez Gene ID
319670
Gene Name
echinoderm microtubule associated protein like 5
Gene Aliases
BC027154, C130068M19Rik
Chromosome Location
Chr. 12: 98786604 - 98901494 on Build GRCm38
UniGene ID
Mm.390484
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081191.1 NP_001074660.1 42 6220
GenBank mRNA AK044024.1 - 6 873
BC027154.1 - 5 765
BC158137.1 - 40 5692 9007401

View Full Product Details