Gene Transcripts

Gene Symbol
Slc41a2
Entrez Gene ID
338365
Gene Name
solute carrier family 41, member 2
Gene Aliases
A230035L05Rik, SLC41A1-L1
Chromosome Location
Chr. 10: 83230848 - 83338329 on Build GRCm38
UniGene ID
Mm.67073
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177388.3 NP_796362.1 2 341
GenBank mRNA AK038549.1 - 2 342
BC138146.1 - 1 179 8861165
BC138147.1 - 1 179 8861166

View Full Product Details