Gene Transcripts

Gene Symbol
Txlng
Entrez Gene ID
353170
Gene Name
taxilin gamma
Gene Aliases
4932441K18Rik, Fiat, Lrp
Chromosome Location
Chr. X: 162778917 - 162829454 on Build GRCm38
UniGene ID
Mm.270186
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001290776.1 NP_001277705.1 11 1446
NM_178935.5 NP_849266.1 10 1593
GenBank mRNA AK030100.1 - 10 1540
AK031023.1 - 8 3132
AK031783.1 - 10 1507
AK044130.1 - 10 1542
BC117898.1 - 11 1387 40096960
BC117899.1 - 11 1387 40096967
DQ320011.1 - 11 1446

View Full Product Details