Gene Transcripts

Gene Symbol
Brsk1
Entrez Gene ID
381979
Gene Name
BR serine/threonine kinase 1
Gene Aliases
Gm1100, SAD-B, SADB
Chromosome Location
Chr. 7: 4690928 - 4715997 on Build GRCm38
UniGene ID
Mm.297064
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001003920.3 NP_001003920.2 12 1360
GenBank mRNA AY533671.1 - 12 1202
BC086636.1 - 12 1359 6402113

View Full Product Details