Gene Transcripts

Gene Symbol
Atxn7l3b
Entrez Gene ID
382423
Gene Name
ataxin 7-like 3B
Gene Aliases
4921506J03Rik, 6230409E21Rik, AI315132, ENSMUSG00000074747
Chromosome Location
Chr. 10: 112925428 - 112929026 on Build GRCm38
UniGene ID
Mm.431239
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033474.2 NP_001028646.2 1 756
GenBank mRNA AK014824.1 - 1 652
BC030168.1 - 1 725
BC047210.1 - 1 726
BC058561.1 - 1 688
BC079579.1 - 1 765
BC094390.1 - 1 714

View Full Product Details