Gene Transcripts
- Gene Symbol
- Igkv4-57-1
- Entrez Gene ID
- 384514
- Gene Name
- immunoglobulin kappa variable 4-57-1
- Gene Aliases
- Gm1418
- Chromosome Location
- Chr. 6: 69544368 - 69544892 on Build GRCm38
- UniGene ID
- Mm.359125
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
J04610.1 |
- |
2 |
88 |
|
View Full Product Details