Gene Transcripts

Gene Symbol
Tmsb15l
Entrez Gene ID
399591
Gene Name
thymosin beta 15b like
Gene Aliases
4930488E11Rik, Tb15r, Tmsb15b1-Tmsb15b2, Tmsb15r
Chromosome Location
Chr. X: 136954988 - 136976874 on Build GRCm38
UniGene ID
Mm.381904
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207267.4 NP_997150.1 3 261
GenBank mRNA AK029707.1 - 3 238
BB623661.1 - 3 257
BC096054.1 - 3 194 3373135
BE625563.1 - 3 186

Gene Symbol
Tmsb15b2
Entrez Gene ID
100034363
Gene Name
thymosin beta 15b2
Gene Aliases
Tb15c
Chromosome Location
Chr. X: 136954988 - 136957979 on Build GRCm38
UniGene ID
Mm.381904
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080967.4 NP_001074436.1 2 121
GenBank mRNA CN836907.1 - 107
CN842750.1 - 2 124

View Full Product Details