Gene Transcripts

Gene Symbol
Eif1
Entrez Gene ID
20918
Gene Name
eukaryotic translation initiation factor 1
Gene Aliases
Sui1-rs1
Chromosome Location
Chr. 11: 100319994 - 100322096 on Build GRCm38
UniGene ID
Mm.13886
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011508.1 NP_035638.1 4 697
GenBank mRNA AK151070.1 - 4 694
AK207284.1 - 1 247
BC003463.1 - 4 697 2648657
BC081429.1 - 4 684 5710951
BC096656.1 - 4 672 5143414
BC134385.1 - 2 360

View Full Product Details