Gene Transcripts

Gene Symbol
Tmem236
Entrez Gene ID
625286
Gene Name
transmembrane protein 236
Gene Aliases
2010003H20Rik, Fam23a, Gm613
Chromosome Location
Chr. 2: 14174524 - 14221993 on Build GRCm38
UniGene ID
Mm.158781
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081310.2 NP_001074779.1 1 142
GenBank mRNA AK144489.1 - 1 143
BY733764.1 - 1 143

View Full Product Details